EMAIL THIS PAGE TO A FRIEND
Journal of biochemistry
Studies on pyruvate kinase (PK) deficiency. II. Electrophoretic, kinetic and immunological studies on pyruvate kinase of erythrocytes and other tissues.
PMID 4781055