EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Quantitative determination of guanethidine and other guanido-containing drugs in biological fluids by gas chromatography with flame ionization detection and multiple ion detection.
PMID 4816566
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91719
Trifluoroacetic anhydride, for GC derivatization, ≥99.0% (GC)
C4F6O3