EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Fluorometric measurement of tyrosine in serum and plasma.
PMID 4818207
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
114693
1-Nitroso-2-naphthol, 97%
C10H7NO2