EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Cytosolic location of an endo-N-acetyl-beta-D-glucosaminidase activity in rat liver and kidney.
PMID 486141
Abstract
Endo-N-acetyl-beta-D-glucosaminidase activity towards an oligomannosidic type glycoamino acid substrate was found in the soluble fraction of rat liver and kidney. No evidence for a lysosomal form of the activity was found.