EMAIL THIS PAGE TO A FRIEND
Journal of electron microscopy
A new embedding method employing D.E.R. 736 and epon 812.
PMID 4875107
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31191 D.E.R. 736