EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Diazoxide and the treatment of hypoglycemia: an historical review.
PMID 4875708
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0950000
Diazoxide, European Pharmacopoeia (EP) Reference Standard
C8H7ClN2O2S