EMAIL THIS PAGE TO A FRIEND
Journal of ultrastructure research
A low-viscosity epoxy resin embedding medium for electron microscopy.
PMID 4887011
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7644
(2-Nonen-1-yl)succinic anhydride, ≥85% (titration)
C13H20O3