EMAIL THIS PAGE TO A FRIEND
The Medical clinics of North America
Bacitracin and tyrothricin.
PMID 4919152
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3000000 Tyrothricin, European Pharmacopoeia (EP) Reference Standard