EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Studies on the metabolism of ATP by isolated bacterial membranes: formation and metabolism of membrane-bound phosphatidic acid.
PMID 4940043