EMAIL THIS PAGE TO A FRIEND
Biochemistry
A membrane-bound phospholipase A1 purified from Escherichia coli.
PMID 4946924
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L3295 Phospholipase A1 from Aspergillus oryzae