EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Pattern of aliphatic dicarboxylic acids in uremic serum including a new organic acid, 2,4-dimethyladipic acid.
PMID 498544
Abstract
(1) 2,4-Dimethyladipic acid was first identified in normal human urine using gas chromatography-mass spectrometry. Urinary excretion of 2,4-dimethyladipic acid in 7 healthy adults ranged from 4.9 mumol to 14 mumol per 24 h. (2) Succinic acid, adipic acid, 3-methyladipic acid, 2,4-dimethyladipic acid, pimelic acid and azelaic acid were identified in the ultrafiltrate of the blood obtained from a chronic uremic patient using a hemodialyzer. (3) Levels of succinic acid, adipic acid, 3-methyladipic acid, 2,4-dimethyladipic acid, pimelic acid and azelaic acid in uremic serum were determined using a mass fragmentographic technique. Concentration of succinic acid in uremic serum was comparable to that in normal serum, whereas concentrations of adipic acid, 3-methyladipic acid, 2,4-dimethyladipic acid, pimelic acid and azelaic acid were highly elevated in uremic serum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P45001
Pimelic acid, 98%
C7H12O4