EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Snake venom toxins. Purification and properties of three toxins from Naja nivea (Linnaeus) (Cape cobra) venom and the amino acid sequence of toxin delta.
PMID 4995745