EMAIL THIS PAGE TO A FRIEND
Biochemistry
Human pancreatic enzymes. Isolation and properties of a major form of chymotrypsin.
PMID 4997645
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8946 α-Chymotrypsin from human pancreas, lyophilized powder