EMAIL THIS PAGE TO A FRIEND
International journal of clinical pharmacology and biopharmacy
The effect of gastro-intestinal filling on the absorption of sulfametoxydiazine.
PMID 500256
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0383
Sulfameter
C11H12N4O3S
46834
Sulfameter, VETRANAL, analytical standard
C11H12N4O3S