EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A new derivative for the gas-liquid chromatographic determination of homovanillic acid.
PMID 5036466
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52517
1,1,1,3,3,3-Hexafluoro-2-propanol, for GC derivatization, LiChropur, ≥99.8%
C3H2F6O