EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
A new chromogenic substrate method for assay of bacterial endotoxins using Limulus hemocyte lysate.
PMID 504254
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76556
Boc-Ser(Bzl)-Gly-Arg-NH-Np.HCl, ≥98.0% (HPLC)
C29H40N8O8 · HCl