EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Galactose utilization in galactosemia.
PMID 5059566
Abstract
Cultures of human galactosemic fibroblasts without detectable transferase activity were able to convert [1-(14)C]galactose to (14)CO(2) to the same extent as normal cells, but did so at a significantly slower rate. The utilization of galactose in both normal and galactosemic cells was strongly inhibited by glucose at physiologic concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
Detroit 510, 90102531