EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
The total synthesis of cephalotaxine.
PMID 5072334
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251356
Methylenecyclopentane, 97%
C6H10