EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Platelet adhesiveness and aggregation. II. Surface sialic acid, glycoprotein: N-acetylneuraminic acid transferase, and neuraminidase of human blood platelets.
PMID 5082512