EMAIL THIS PAGE TO A FRIEND
Journal of molecular biology
Selective dissociation of histones from chromatin by sodium deoxycholate.
PMID 5104701
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30970
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
C24H39NaO4