EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Fluorescence and phosphorescence of 5- and 8-aminoquinoline.
PMID 5129575
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
260789
8-Aminoquinoline, 98%
C9H8N2