EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Cellular immunity to renal carcinomas in man.
PMID 5137312
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
T24/83, 85061107