EMAIL THIS PAGE TO A FRIEND
Nature
Epsilon-N-dimethyllysine in amoeba actin.
PMID 5212577
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19773
Nε,Nε-Dimethyl-L-lysine monohydrochloride, ≥96% (TLC)
C8H18N2O2 · HCl