EMAIL THIS PAGE TO A FRIEND
Comptes-rendus des travaux du Laboratoire Carlsberg
A review on prorennin and rennin.
PMID 5330666
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R4877 Rennin from calf stomach, ≥20 units/mg protein