EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
Chromosomes of 14 hematopoietic cell lines derived from peripheral blood of persons with and without chromosome anomalies.
PMID 5353241
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
RPMI 1788