EMAIL THIS PAGE TO A FRIEND
The American journal of anatomy
Rib development in chick embryos analyzed by means of tantalum foil blocks.
PMID 5362271
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF76534500
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Ta
GF36069598
Tantalum, foil, 100x100mm, thickness 0.75mm, annealed, 99.9%
Ta
GF35827252
Tantalum, foil, 50x50mm, thickness 2.0mm, 99.999%
Ta
GF80276182
Tantalum, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Ta
GF85784653
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Ta
GF65369324
Tantalum, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.9%
Ta
GF14275852
Tantalum, foil, 25x25mm, thickness 0.25mm, annealed, 99.9%
Ta
GF17240554
Tantalum, foil, 25mm disks, thickness 0.006mm, 99.9%
Ta
GF65422739
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, flattened, 99.9%
Ta
GF10417698
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF77910082
Tantalum, foil, light tested, 100x100mm, thickness 0.015mm, 99.9%
Ta
GF68984229
Tantalum, foil, 100x100mm, thickness 1.0mm, annealed, 99.9%
Ta
GF18537925
Tantalum, foil, 15mm disks, thickness 0.006mm, 99.9%
Ta
GF43143072
Tantalum, foil, light tested, 50x50mm, thickness 0.02mm, 99.9%
Ta
GF19254472
Tantalum, foil, not light tested, 25x25mm, thickness 0.0005mm, 99.9%
Ta
GF34522703
Tantalum, foil, 150x150mm, thickness 0.125mm, annealed, 99.9%
Ta
GF66812564
Tantalum, foil, 150x150mm, thickness 0.100mm, annealed, 99.9%
Ta
GF86342485
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Ta
GF75067702
Tantalum, foil, 150x150mm, thickness 0.15mm, annealed, flattened, 99.9%
Ta
GF98447103
Tantalum, foil, 25x25mm, thickness 0.75mm, annealed, 99.9%
Ta
GF60298195
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.9%
Ta
GF10710593
Tantalum, foil, 25x25mm, thickness 2.0mm, annealed, 99.9%
Ta
GF15877132
Tantalum, foil, 25x25mm, thickness 0.125mm, annealed, 99.9%
Ta
GF34817474
Tantalum, foil, light tested, 120x120mm, thickness 0.025mm, as rolled, 99.9%
Ta
GF84642206
Tantalum, foil, 50x50mm, thickness 0.125mm, annealed, 99.9%
Ta
GF49190249
Tantalum, foil, 50x50mm, thickness 0.5mm, annealed, 99.9%
Ta
GF19196310
Tantalum, foil, 1m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF41727832
Tantalum, foil, light tested, 25x25mm, thickness 0.075mm, annealed, 99.9%
Ta
GF34465888
Tantalum, foil, 0.5m coil, thickness 0.034mm, as rolled, 99.9%
Ta
GF96084603
Tantalum, foil, 0.5m coil, thickness 0.03mm, as rolled, 99.9%
Ta