EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Superoxide dismutase. An enzymic function for erythrocuprein (hemocuprein).
PMID 5389100
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X0626
Xanthine, ≥99.5% (HPLC), purified by recrystallization
C5H4N4O2
X7375
Xanthine, ≥99%
C5H4N4O2