EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
New substrate for galactose oxidase.
PMID 5413941
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D107204
1,3-Dihydroxyacetone dimer, 97%
C6H12O6