EMAIL THIS PAGE TO A FRIEND
Cancer research
Transplantable rat mammary tumors induced by 5-nitro-2-furaldehyde semicarbazone and by formic acid 2-[4-(5-nitro-2-furyl)-2-thiazolyl] hydrazide.
PMID 5426943
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73340
5-Nitro-2-furaldehyde semicarbazone, ≥97.0% (HPLC)
C6H6N4O4