EMAIL THIS PAGE TO A FRIEND
Toxicology
Synergistic effects on the toxicity of organotins on cotton leafworms.
PMID 542951
Abstract
Piperonyl butoxide, sulfoxide and MGK 264 were found to act as synergists for certain organotin compounds; with respect to their toxicity on the larvae of a susceptible strain of the cotton leafworm, Spodoptera littoralis (Boisd.). High degree of synergism was obtained when either piperonyl butoxide or MGK 264 was combined with Plictran and/or MGK 264 when combined with Duter. The 3 synergists were equivalent in their synergistic action with Brestan. In all cases, the relatively high degree of synergism was brought about when organotins and synergists were mixed at ratio of 10 : 1. On the basis of synergistic ratios, data indicated that detoxification mechanism of Plictran may be more sensitive to synergistic effects than that of Duter and Brestan. Testing organotin compounds against the more tolerable field strain provided probable evidence of negative correlation between different insecticides and organotins, especially Plictran.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36168
MGK 264, PESTANAL®, analytical standard
C17H25NO2