EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Coexistence of different pathways in the metabolism of n-propylbenzene by Pseudomonas sp.
PMID 543699
Abstract
Pseudomonas desmolytica S449B1 and Pseudomonas convexa S107B1 grown on n-propylbenzene oxidized n-propylbenzene to beta-phenylpropionic acid and benzoic acid by initial oxidation of the n-propyl side chain and the following beta-oxidation, respectively. The same strains also oxidized n-propylbenzene to 3-n-propylcatechol by initial oxidation of positions 2 and 3 of the aromatic nucleus. A ring fission product, 2-hydroxy-6-oxononanoic acid, was also isolated from the culture broth. Together with the results of oxygen uptake experiments, the data obtained suggested not only the existence of a reductive step to form 2-hydroxy-6-oxononanoic acid, but also the coexistence of two different pathways in the metabolism of n-propylbenzene by the strains used.