EMAIL THIS PAGE TO A FRIEND
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences
[Hygienic chemical studies on plastics (II). Residual acrylonitrile in ABS resin or AS resin and its migration into water (author's transl)].
PMID 543964