EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Sequence analysis of microgram amounts of peptides by mass spectrometry.
PMID 5440913
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33085
Acetic anhydride
C4H6O3