EMAIL THIS PAGE TO A FRIEND
Archivos de farmacologia y toxicologia
[Anorexigenic activity of various derivatives of alpha-aminoacetophenone].
PMID 546333
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A38207
2-Aminoacetophenone hydrochloride, 99%
C8H9NO · HCl