EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Isolation and identification of N-G,N-G- and N-G,N'-G-dimethyl-arginine, N-epsilon-mono-, di-, and trimethyllysine, and glucosylgalactosyl- and galactosyl-delta-hydroxylysine from human urine.
PMID 5472370
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19773
Nε,Nε-Dimethyl-L-lysine monohydrochloride, ≥96% (TLC)
C8H18N2O2 · HCl