EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Antispasmodic agents. 1. Syntheses and pharmacological activity of aminoalkyl 3-substituted phenylacetates.
PMID 5539757
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209392
3-Methoxyphenylacetonitrile, 99%
C9H9NO