EMAIL THIS PAGE TO A FRIEND
West African journal of pharmacology and drug research
The metabolism of 2-naphthylacetic acid in cat and rhesus monkey.
PMID 555134
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4002
2-Naphthaleneacetic acid, 99%
C12H10O2