EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
GLC-mass spectral analysis of psilocin and psilocybin.
PMID 559753
Abstract
With the combined technique of GLC-mass spectrometry, psilocin and psilocybin, two hallucinogenic indoles, were analyzed as their trimethylsilyl derivatives. The method was applied to these two components in an extract of Psilocybe cubensis (Earle) Sing.