EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Purification, crystallization and properties of the D-xylose isomerase from Lactobacillus brevis.
PMID 5646045
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4166 Glucose Isomerase from Streptomyces murinus, ≥350 U/g