EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Purification and properties of phospholipase A-1 from rat and calf brain.
PMID 5657461
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L3295 Phospholipase A1 from Thermomyces lanuginosus