EMAIL THIS PAGE TO A FRIEND
Experientia
Peroxidative activity of hemepeptides from horse heart cytochrome c.
PMID 5661404
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6756 Microperoxidase (MP-11) sodium salt, ≥85% (HPLC)
C84H116FeN20O21S2