EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Reaction of 2-chlorobenzoxazole with anthranilic acids and 2-amino-1-naphthalenesulfonic acid.
PMID 5677361
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
291137
2-Amino-1-naphthalenesulfonic acid, 98%
C10H9NO3S
274089
2-Chlorobenzoxazole, 99%
C7H4ClNO