EMAIL THIS PAGE TO A FRIEND
Biochemistry
Sulfenyl halides as modifying reagents for polypeptides and proteins. 3. Azobenzene-2-sulfenyl bromide, a selective reagent for cysteinyl residues.
PMID 5722259