EMAIL THIS PAGE TO A FRIEND
Indian journal of physiology and pharmacology
Certain aspects of pharmacological profiles of chlordiazepoxide and diazepam.
PMID 5737357
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
SV-T2, 90030801