EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The mechanism of inactivation of glyceraldehyde 3-phosphate dehydrogenase by tetrathionate, o-iodosobenzoate, and iodine monochloride.
PMID 5773281
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72030
Sodium tetrathionate dihydrate, purum p.a., ≥98.0% (T)
Na2O6S4 · 2H2O