EMAIL THIS PAGE TO A FRIEND
Journal of the Chemical Society. Perkin transactions 1
Oxidation and Claisen condensation products of 3-nitro-o-xylene.
PMID 5817990
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130303
1,2-Dimethyl-3-nitrobenzene, 97%
C8H9NO2