EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Preparation of glycosyl phosphates. Betad-fructopyranose 2-phosphate.
PMID 5902475
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44627
D-Fructopyranose 2-phosphate potassium salt, ≥98.0% (TLC)
C6H13O9P · xK+