EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The biosynthesis of cell wall lipopolysaccharide in Escherichia coli. V. Purification and properties of 3-deoxy-D-manno-octulosonate aldolase.
PMID 5912115
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
67891 3-Deoxy-D-manno-octulosonate Aldolase from E. coli, ≥1.0 U/mg