EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Aromatization of cyclohexanecarboxylic acid.
PMID 5916383
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
101834
Cyclohexanecarboxylic acid, 98%
C7H12O2
29252
Cyclohexene-1-carbonitrile, ≥97.0%
C7H9N