EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Mechanism of action of guinea pig liver transglutaminase. I. Purification and properties of the enzyme: identification of a functional cysteine essential for activity.
PMID 5928192